Assay Details
Gene Symbol: Ldhc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013580.4 | NP_038608.1 | 1 - 2 | 40 | 63 | |
GenBank mRNA | AK007058.1 | - | 1 - 2 | 41 | 63 | |
AK161399.1 | - | 1 - 2 | 28 | 63 | ||
BC049602.1 | - | 1 - 2 | 44 | 6774010 | 63 | |
X04752.1 | - | 1 - 2 | 48 | 63 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK007058.1,AK161399.1,BC049602.1,NM_013580.4,X04752.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
