Assay Details
Gene Symbol: Lrrc23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK015011.1 | - | 2 - 3 | 207 | 96 | |
BC049601.1 | - | 2 - 3 | 214 | 6743951 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
