Assay Details
| Gene Symbol: Lrrc23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015011.1 | - | 3 - 4 | 315 | 74 | |
| BC049601.1 | - | 3 - 4 | 322 | 6743951 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lrrc23
Genomic Map