Assay Details
Gene Symbol: Lrrc23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC049601.1 | - | 4 - 5 | 578 | 6743951 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1960411 | AK015011.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1960411 | BC049601.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1960411 | NM_013588.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 405008 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 416044 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 22451 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 970937 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 767186 | AK041830.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 26532 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P145167 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1960411 | AK015011.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1960411 | BC049601.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1960411 | NM_013588.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816971 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816987 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816935 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816949 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816945 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816956 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816951 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816990 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816995 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816983 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816978 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816938 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816946 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816982 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816993 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816976 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816980 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816953 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816985 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816939 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816937 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816996 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816958 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816954 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816991 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816973 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816968 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816975 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816942 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816972 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816941 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816955 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06056334 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P145167 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01437722 | - | - |
Whole Mouse Genome G4122A | A_51_P489679 | BC049601.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P489679 | NM_013588.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118480 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P145167 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05589527 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05547804 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05396797 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05547796 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05547805 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05547795 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05589526 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05589514 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05589518 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816949 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816873 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816914 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816954 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816925 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816997 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816901 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816903 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816982 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816917 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816945 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816922 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816990 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816893 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816887 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816912 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816942 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816896 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816985 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816987 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816929 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816918 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816915 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816883 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816895 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816964 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816975 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816880 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816889 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816924 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816909 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816920 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817002 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816890 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816999 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816941 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816907 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816927 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816978 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816933 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816888 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816961 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816905 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816871 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817000 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816879 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816955 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816972 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816993 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816877 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816866 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816937 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817007 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816932 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816935 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816875 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816980 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816976 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816983 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816939 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816953 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816892 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816965 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816872 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816910 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816998 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816870 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816885 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816971 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816947 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816974 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816946 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816962 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24816976 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24816965 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24816937 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24816985 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24816962 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24816974 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05396797 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24816961 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24817001 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05026903 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24817002 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05370614 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24816999 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05681342 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24816980 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24816939 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24816967 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24816969 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05779255 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05370617 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05589522 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24817005 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24816964 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24816935 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24817007 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24817000 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24816987 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24816946 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05248147 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24816996 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24816982 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24816945 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24816972 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24816953 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05529247 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24816997 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24816959 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24816983 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24816991 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24816990 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24816941 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24816975 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24816947 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24816949 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24817004 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24816978 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05819953 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24816958 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24816994 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10547823 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10547822 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10547825 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10547824 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10547829 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10547827 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10547828 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10547821 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10547826 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1432083_PM_a_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10547820 | - | - |
Mouse Genome 430 2.0 Array | 1432083_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1432083_a_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16977_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16977_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0600024202.mm.1 | TC0600003117.mm.1 | MTA TC |
Murine Genome U74v2 Set | 161392_PM_f_at | - | - |
Murine Genome U74v2 Set | 161392_f_at | - | - |
Murine Genome U74v2 Set | 104667_at | - | - |
Murine Genome U74v2 Set | 104667_PM_at | - | - |
Murine Genome U74v2 Set | 169586_PM_i_at | - | - |
Murine Genome U74v2 Set | 169586_i_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1432083_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1432083_a_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 923656 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 3130066 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6370504 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003130066 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006370504 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1257820 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1228163 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1228163 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1257820 | - | - |
mouse-6 v1.1 (Illumina) | scl28421.8.141_12-S | - | - |
mouse-6 v1.1 (Illumina) | scl0001258.1_137-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6370504 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3130066 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl28421.8.141_12-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0001258.1_137-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6370504 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3130066 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1228163 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1257820 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1228163 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1257820 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl28421.8.141_12-S | - | - |
