Assay Details
| Gene Symbol: Tcp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013686.4 | NP_038714.2 | 2 - 3 | 387 | 84 | |
| GenBank mRNA | AK149611.1 | - | 2 - 3 | 252 | 84 | |
| AK149755.1 | - | 2 - 3 | 253 | 84 | ||
| AK150666.1 | - | 2 - 3 | 267 | 84 | ||
| AK150744.1 | - | 2 - 3 | 267 | 84 | ||
| AK151068.1 | - | 2 - 3 | 252 | 84 | ||
| AK151903.1 | - | 2 - 3 | 267 | 84 | ||
| AK152633.1 | - | 2 - 3 | 267 | 84 | ||
| AK152641.1 | - | 2 - 3 | 244 | 84 | ||
| AK152759.1 | - | 2 - 3 | 253 | 84 | ||
| AK153037.1 | - | 2 - 3 | 253 | 84 | ||
| AK165665.1 | - | 2 - 3 | 254 | 84 | ||
| AK166828.1 | - | 2 - 3 | 251 | 84 | ||
| AK166966.1 | - | 2 - 3 | 275 | 84 | ||
| AK167529.1 | - | 2 - 3 | 252 | 84 | ||
| BC003809.1 | - | 2 - 3 | 235 | 3599670 | 84 | |
| D90344.1 | - | 2 - 3 | 202 | 84 | ||
| M12899.1 | - | 2 - 3 | 210 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tcp1
Genomic Map