Assay Details
Gene Symbol: Jag1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013822.5 | NP_038850.1 | 16 - 17 | 2379 | 94 | |
GenBank mRNA | AF171092.1 | - | 16 - 17 | 2384 | 94 | |
AK137092.1 | - | 16 - 17 | 2279 | 94 | ||
BC058675.1 | - | 16 - 17 | 2191 | 6834418 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
