Assay Details
| Gene Symbol: Ddb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015735.1 | NP_056550.1 | 18 - 19 | 2356 | 88 | |
| GenBank mRNA | AB026432.1 | - | 18 - 19 | 2356 | 88 | |
| AF159853.1 | - | 18 - 19 | 2316 | 88 | ||
| AK145328.1 | - | 6 - 7 | 654 | 88 | ||
| AK146522.1 | - | 18 - 19 | 2414 | 88 | ||
| AK154303.1 | - | 18 - 19 | 2382 | 88 | ||
| AK155020.1 | - | 18 - 19 | 2437 | 88 | ||
| AK155920.1 | - | 18 - 19 | 2397 | 88 | ||
| AK157491.1 | - | 18 - 19 | 2436 | 88 | ||
| BC009661.1 | - | 18 - 19 | 2363 | 3487617 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ddb1
Genomic Map