Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200029746.mm.1 |
NM_001289521.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029746.mm.1 |
TC0200003473.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200029730.mm.1 |
NM_001289521.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029730.mm.1 |
NM_015753.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029730.mm.1 |
TC0200003473.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200029727.mm.1 |
NM_001289521.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029727.mm.1 |
NM_015753.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029727.mm.1 |
TC0200003473.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200029745.mm.1 |
NM_001289521.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029745.mm.1 |
TC0200003473.mm.1 |
MTA TC |