Assay Details
| Gene Symbol: Uba2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016682.2 | NP_057891.1 | 5 - 6 | 479 | 64 | |
| GenBank mRNA | AK146925.1 | - | 5 - 6 | 525 | 64 | |
| AK151765.1 | - | 5 - 6 | 484 | 64 | ||
| AK152415.1 | - | 5 - 6 | 503 | 64 | ||
| AK152977.1 | - | 5 - 6 | 484 | 64 | ||
| AK163451.1 | - | 5 - 6 | 475 | 64 | ||
| AK164826.1 | - | 5 - 6 | 495 | 64 | ||
| AK165321.1 | - | 5 - 6 | 501 | 64 | ||
| AK166133.1 | - | 5 - 6 | 511 | 64 | ||
| AK168673.1 | - | 5 - 6 | 491 | 64 | ||
| AK169168.1 | - | 5 - 6 | 475 | 64 | ||
| AK182052.1 | - | 1 - 2 | 44 | 64 | ||
| BC054768.1 | - | 5 - 6 | 505 | 6415071 | 64 | |
| U35833.1 | - | 5 - 6 | 480 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Uba2
Genomic Map