Assay Details
| Gene Symbol: Uba2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016682.2 | NP_057891.1 | 10 - 11 | 1052 | 98 | |
| GenBank mRNA | AK146925.1 | - | 10 - 11 | 1098 | 98 | |
| AK151765.1 | - | 10 - 11 | 1057 | 98 | ||
| AK152415.1 | - | 10 - 11 | 1076 | 98 | ||
| AK152977.1 | - | 10 - 11 | 1058 | 98 | ||
| AK163451.1 | - | 10 - 11 | 1048 | 98 | ||
| AK164826.1 | - | 10 - 11 | 1068 | 98 | ||
| AK165321.1 | - | 10 - 11 | 1074 | 98 | ||
| AK166133.1 | - | 10 - 11 | 1084 | 98 | ||
| AK168673.1 | - | 10 - 11 | 1064 | 98 | ||
| AK169168.1 | - | 10 - 11 | 1048 | 98 | ||
| BC054768.1 | - | 10 - 11 | 1078 | 6415071 | 98 | |
| U35833.1 | - | 10 - 11 | 1053 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Uba2
Genomic Map