Assay Details
| Gene Symbol: Mzf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290453.1 | NP_001277382.1 | 2 - 3 | 723 | 79 | |
| NM_145819.2 | NP_665818.2 | 2 - 3 | 723 | 79 | ||
| GenBank mRNA | AB007407.1 | - | 1 - 2 | 942 | 79 | |
| AB088478.1 | - | 2 - 3 | 624 | 79 | ||
| AK030410.1 | - | 2 - 3 | 724 | 79 | ||
| BC060649.1 | - | 2 - 3 | 683 | 79 | ||
| BC129905.1 | - | 2 - 3 | 473 | 40111655 | 79 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001290453.1 transcripts.
Set Membership: |
|

Mzf1
Genomic Map