Assay Details
| Gene Symbol: Gbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289493.1 | NP_001276422.1 | 1 - 2 | 192 | 63 | |
| GenBank mRNA | U44731.1 | - | 1 - 2 | 81 | 63 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001289493.1, U44731.1 transcripts.
Set Membership: |
|

Gbp3
Genomic Map