Assay Details
Gene Symbol: Gbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289492.1 | NP_001276421.1 | 7 - 8 | 1252 | 79 | |
NM_001289493.1 | NP_001276422.1 | 6 - 7 | 1077 | 79 | ||
NM_018734.3 | NP_061204.3 | 6 - 7 | 1114 | 79 | ||
GenBank mRNA | AK132266.1 | - | 6 - 7 | 1113 | 79 | |
BC019195.1 | - | 7 - 8 | 1163 | 5036920 | 79 | |
U44731.1 | - | 6 - 7 | 966 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2472435 | U44731.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2472435 | AK132266.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2472435 | BC019195.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2472435 | NM_018734.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2472435 | AA162247.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 107643 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 562140 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 380340 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 66359 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 12709 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 264175 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 750035 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 379352 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 82527 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P114154 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P159064 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2472435 | U44731.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2472435 | AK132266.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2472435 | BC019195.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2472435 | NM_018734.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2472435 | AA162247.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P165244 | U44731.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P165244 | AK132266.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P165244 | NM_018734.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P165244 | AA162247.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22591505 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22591481 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22591495 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22591500 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22591510 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22591504 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22591485 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22591508 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22591519 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22591496 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22591494 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22591499 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22591507 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22591509 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22591515 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22591502 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22591513 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22591486 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22591493 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22591517 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22591498 | - | - |
Whole Mouse Genome G4122A | A_51_P165244 | U44731.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P165244 | AK132266.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P165244 | NM_018734.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P165244 | AA162247.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101179 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02749895 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03120801 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02749902 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02749900 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03120797 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02749901 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02749897 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02548729 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591543 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591527 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591542 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591582 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591549 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591597 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591547 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591583 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591499 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591505 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591525 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591609 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591464 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591509 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591532 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591461 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591579 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591611 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591554 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591566 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591563 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591537 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591571 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591605 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591481 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591533 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591596 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591539 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591567 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591538 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591468 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591588 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591565 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591595 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591544 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591494 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591556 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591493 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591545 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591502 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591592 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591473 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591529 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591469 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591521 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591617 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591552 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591513 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591535 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591500 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591467 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591536 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591562 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591568 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591476 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591524 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591504 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591510 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591520 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591613 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591580 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591507 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591479 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591515 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591498 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591618 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591519 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591577 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591590 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591496 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591593 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591523 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591573 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591604 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22591559 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591529 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591467 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02360751 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591510 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591476 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591482 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591556 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02548727 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591479 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591500 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591542 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591493 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591519 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591515 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591475 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591502 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591532 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03123510 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591462 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591533 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591494 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591523 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591535 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591505 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591525 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591562 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03190704 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591461 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591537 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591536 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591559 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591463 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591507 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591498 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591513 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591528 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02360744 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591544 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591524 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591481 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591504 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591495 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02749901 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591545 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03120798 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591552 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591543 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591539 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591520 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591547 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02748540 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591496 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03190700 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591554 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591538 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22591486 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10496586 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10496583 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10496590 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10496591 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10496589 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10496587 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10496588 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10496581 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10496582 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10496584 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10496585 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1418392_PM_a_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10496580 | - | - |
Mouse Genome 430 2.0 Array | 1418392_a_at | - | - |
Mouse Genome 430 2.0 Array | 1418392_PM_a_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 55932_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 55932_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0300011664.mm.1 | NM_001289492.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300011664.mm.1 | NM_001289493.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300011664.mm.1 | NM_018734.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300011664.mm.1 | NM_018734.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300011664.mm.1 | TC0300001446.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300011663.mm.1 | NM_001289492.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300011663.mm.1 | NM_001289493.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300011663.mm.1 | NM_018734.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300011663.mm.1 | NM_018734.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300011663.mm.1 | TC0300001446.mm.1 | MTA TC |
Mouse Expression Set 430 | 1418392_PM_a_at | - | - |
Mouse Expression Set 430 | 1418392_a_at | - | - |
Murine Genome U74v2 Set | 103202_at | - | - |
Murine Genome U74v2 Set | 103202_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1418392_PM_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1418392_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1418392_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1418392_a_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 405120 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 6520471 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6520471 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006520471 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1244513 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1244513 | - | - |
mouse-6 v1.1 (Illumina) | scl22518.14.1_14-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6520471 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl22518.14.1_14-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6520471 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2918002 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1244513 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1244513 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2918002 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl22518.14.1_14-S | - | - |
