Assay Details
| Gene Symbol: Igfbpl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018741.2 | NP_061211.1 | 2 - 3 | 583 | 74 | |
| GenBank mRNA | AB006141.1 | - | 2 - 3 | 571 | 74 | |
| AK012705.1 | - | 2 - 3 | 584 | 75 | ||
| BC051960.1 | - | 2 - 3 | 553 | 5710646 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Igfbpl1
Genomic Map