Assay Details
| Gene Symbol: Agpat6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018743.4 | NP_061213.2 | 9 - 10 | 1937 | 76 | |
| GenBank mRNA | AF015811.1 | - | 6 - 7 | 613 | 76 | |
| AF406611.1 | - | 8 - 9 | 1448 | 76 | ||
| AK045235.1 | - | 9 - 10 | 1831 | 76 | ||
| AK077561.1 | - | 9 - 10 | 1821 | 76 | ||
| AK083589.1 | - | 9 - 10 | 1831 | 76 | ||
| AK161270.1 | - | 9 - 10 | 1800 | 76 | ||
| AK162033.1 | - | 6 - 7 | 742 | 76 | ||
| AK167876.1 | - | 9 - 10 | 1846 | 76 | ||
| AK169257.1 | - | 9 - 10 | 1831 | 76 | ||
| AK184759.1 | - | 3 - 4 | 273 | 76 | ||
| AK199339.1 | - | 3 - 4 | 226 | 76 | ||
| AK203007.1 | - | 3 - 4 | 212 | 76 | ||
| AK216721.1 | - | 3 - 4 | 273 | 76 | ||
| AK218609.1 | - | 3 - 4 | 253 | 76 | ||
| AY489184.1 | - | 8 - 9 | 1006 | 76 | ||
| BC031767.1 | - | 8 - 9 | 1291 | 4457846 | 76 | |
| BC057860.1 | - | 6 - 7 | 611 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Agpat6
Genomic Map