Assay Details
| Gene Symbol: Slc23a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018824.2 | NP_061294.2 | 9 - 10 | 1182 | 64 | |
| GenBank mRNA | AB038145.1 | - | 8 - 9 | 918 | 64 | |
| AY004874.1 | - | 9 - 10 | 1136 | 64 | ||
| BC050823.1 | - | 8 - 9 | 946 | 6332451 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc23a2
Genomic Map