Assay Details
Gene Symbol: Impa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018864.5 | NP_061352.2 | 3 - 4 | 319 | 111 | |
GenBank mRNA | AF042730.1 | - | 3 - 4 | 264 | 111 | |
AK011770.1 | - | 3 - 4 | 257 | 111 | ||
AK078406.1 | - | 3 - 4 | 268 | 111 | ||
AK083238.1 | - | 3 - 4 | 253 | 111 | ||
AK145472.1 | - | 256 | 111 | |||
AK147778.1 | - | 3 - 4 | 279 | 111 | ||
AK152774.1 | - | 3 - 4 | 270 | 111 | ||
AK153457.1 | - | 3 - 4 | 286 | 111 | ||
AK160203.1 | - | 3 - 4 | 257 | 111 | ||
AK160467.1 | - | 3 - 4 | 251 | 111 | ||
AK165977.1 | - | 3 - 4 | 261 | 111 | ||
AK168457.1 | - | 3 - 4 | 294 | 111 | ||
BC049080.1 | - | 3 - 4 | 260 | 6419835 | 111 | |
BC055722.1 | - | 2 - 3 | 214 | 6413389 | 111 | |
BY283562.1 | - | 3 - 4 | 320 | 111 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
