Assay Details
| Gene Symbol: Lmna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002011.3 | NP_001002011.2 | 5 - 6 | 1187 | 83 | |
| NM_001111102.2 | NP_001104572.1 | 5 - 6 | 1187 | 83 | ||
| NM_019390.3 | NP_062263.1 | 5 - 6 | 658 | 83 | ||
| GenBank mRNA | AK004619.2 | - | 5 - 6 | 1142 | 83 | |
| AK147150.1 | - | 5 - 6 | 1143 | 83 | ||
| AK149998.1 | - | 5 - 6 | 1143 | 83 | ||
| AK150391.1 | - | 5 - 6 | 1143 | 83 | ||
| AK150501.1 | - | 5 - 6 | 1143 | 83 | ||
| AK150624.1 | - | 5 - 6 | 1143 | 83 | ||
| AK152539.1 | - | 5 - 6 | 1143 | 83 | ||
| AK152646.1 | - | 5 - 6 | 1143 | 83 | ||
| AK152846.1 | - | 5 - 6 | 1143 | 83 | ||
| AK153295.1 | - | 5 - 6 | 1084 | 83 | ||
| AK161221.1 | - | 5 - 6 | 1008 | 83 | ||
| AK167858.1 | - | 5 - 6 | 1140 | 83 | ||
| AK215823.1 | - | 2 - 3 | 266 | 83 | ||
| BC015302.1 | - | 5 - 6 | 1063 | 4240057 | 83 | |
| BC094020.1 | - | 5 - 6 | 1049 | 4187374 | 83 | |
| D13181.1 | - | 2 - 3 | 236 | 83 | ||
| D14850.1 | - | 5 - 6 | 658 | 83 | ||
| DQ832702.1 | - | 5 - 6 | 938 | 83 | ||
| DQ832703.1 | - | 5 - 6 | 938 | 83 | ||
| X14170.1 | - | 5 - 6 | 1132 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lmna
Genomic Map