Assay Details
| Gene Symbol: Lmna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002011.3 | NP_001002011.2 | 8 - 9 | 1735 | 63 | |
| NM_001111102.2 | NP_001104572.1 | 8 - 9 | 1735 | 63 | ||
| NM_019390.3 | NP_062263.1 | 8 - 9 | 1206 | 63 | ||
| GenBank mRNA | AK003174.1 | - | 1 - 2 | 86 | 63 | |
| AK004619.2 | - | 8 - 9 | 1690 | 63 | ||
| AK147150.1 | - | 8 - 9 | 1691 | 63 | ||
| AK149998.1 | - | 8 - 9 | 1691 | 63 | ||
| AK150391.1 | - | 8 - 9 | 1691 | 63 | ||
| AK150501.1 | - | 8 - 9 | 1691 | 63 | ||
| AK150624.1 | - | 8 - 9 | 1691 | 63 | ||
| AK152539.1 | - | 8 - 9 | 1691 | 63 | ||
| AK152646.1 | - | 8 - 9 | 1691 | 63 | ||
| AK152846.1 | - | 8 - 9 | 1691 | 63 | ||
| AK153295.1 | - | 8 - 9 | 1632 | 63 | ||
| AK161221.1 | - | 8 - 9 | 1556 | 63 | ||
| AK167858.1 | - | 8 - 9 | 1688 | 63 | ||
| AK185150.1 | - | 2 - 3 | 129 | 63 | ||
| AK208885.1 | - | 2 - 3 | 130 | 63 | ||
| AK216120.1 | - | 2 - 3 | 130 | 63 | ||
| BC015302.1 | - | 8 - 9 | 1611 | 4240057 | 63 | |
| BC094020.1 | - | 8 - 9 | 1597 | 4187374 | 63 | |
| D13181.1 | - | 5 - 6 | 784 | 63 | ||
| D14850.1 | - | 8 - 9 | 1206 | 63 | ||
| DQ832702.1 | - | 8 - 9 | 1486 | 63 | ||
| DQ832703.1 | - | 8 - 9 | 1486 | 63 | ||
| X14170.1 | - | 8 - 9 | 1680 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lmna
Genomic Map