Assay Details
Gene Symbol: Lrrc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019457.2 | NP_062330.1 | 1 - 2 | 133 | 70 | |
GenBank mRNA | AF092208.1 | - | 1 - 2 | 78 | 70 | |
AK016236.1 | - | 1 - 2 | 122 | 70 | ||
AK077124.1 | - | 1 - 2 | 117 | 70 | ||
BC046277.1 | - | 1 - 2 | 84 | 6310345 | 70 | |
CJ046721.1 | - | 1 - 2 | 134 | 70 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AF092208.1,AK016236.1,AK077124.1,BC046277.1,NM_019457.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
