Assay Details
| Gene Symbol: Olfr157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019475.3 | NP_062348.1 | 1 | 1550 | 75 | |
| GenBank mRNA | BC137911.1 | - | 1 | 1547 | 8860930 | 75 |
| BC137912.1 | - | 1 | 1547 | 8860931 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P504090 | BC137912.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P504090 | NM_019475.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P504090 | BC137911.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P504090 | BC137912.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P504090 | NM_019475.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P504090 | BC137911.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2169004 | BC137912.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2169004 | NM_019475.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2169004 | BC137911.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 839414 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P166784 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P504090 | BC137912.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P504090 | NM_019475.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P504090 | BC137911.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P504090 | BC137912.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P504090 | NM_019475.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P504090 | BC137911.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2169004 | BC137912.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2169004 | NM_019475.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2169004 | BC137911.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P504090 | BC137912.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P504090 | NM_019475.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P504090 | BC137911.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P504090 | BC137912.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P504090 | NM_019475.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P504090 | BC137911.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22873871 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22873894 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22873900 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22873909 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22873904 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22873867 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22873899 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22873885 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22873910 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22873890 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22873903 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22873896 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22873898 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22873902 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22873873 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22873906 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22873908 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22873874 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22873892 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22873868 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22873888 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22873886 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P166784 | - | - |
| Whole Mouse Genome G4122A | A_51_P504090 | BC137912.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P504090 | NM_019475.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P504090 | BC137911.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P504090 | BC137912.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P504090 | NM_019475.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P504090 | BC137911.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120099 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P166784 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03812687 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03309828 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03827000 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03827001 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03812678 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03309826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873885 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03851992 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22873895 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22873859 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22873868 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22873898 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22873894 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03228606 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03452907 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22873872 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22873890 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22873888 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03452908 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22873864 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22873886 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22873911 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03812687 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22873883 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03284581 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22873896 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22873854 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22873873 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03284587 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03505540 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03505543 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22873874 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03309828 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22873862 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22873885 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22873902 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22873910 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22873900 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22873892 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22873906 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22873856 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22873866 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03440950 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10512638 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10512639 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10512631 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1460312_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10512637 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10512630 | - | - |
| Mouse Genome 430 2.0 Array | 1460312_at | - | - |
| Mouse Genome 430 2.0 Array | 1460312_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400020778.mm.1 | TC0400002546.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400020778.mm.1 | NM_019475.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400020778.mm.1 | NM_019475.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400020780.mm.1 | TC0400002546.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400020780.mm.1 | NM_019475.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400020780.mm.1 | NM_019475.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400020781.mm.1 | TC0400002546.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400020781.mm.1 | NM_019475.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400020781.mm.1 | NM_019475.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400020779.mm.1 | TC0400002546.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400020779.mm.1 | NM_019475.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400020779.mm.1 | NM_019475.3 | Refseq |
| Mouse Expression Set 430 | 1460312_at | - | - |
| Mouse Expression Set 430 | 1460312_PM_at | - | - |
| Murine Genome U74v2 Set | 97001_r_at | - | - |
| Murine Genome U74v2 Set | 97000_s_at | - | - |
| Murine Genome U74v2 Set | 97000_PM_s_at | - | - |
| Murine Genome U74v2 Set | 97001_PM_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460312_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460312_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1460312_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1460312_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 493685 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2320110 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002320110 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1229628 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1229628 | - | - |
| mouse-6 v1.1 (Illumina) | scl24389.1.2_18-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2320110 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl24389.1.2_18-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2320110 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1229628 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1229628 | - | - |

Olfr157
Genomic Map