Assay Details
| Gene Symbol: Uchl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159866.1 | NP_001153338.1 | 2 - 3 | 262 | 96 | |
| NM_019562.2 | NP_062508.2 | 2 - 3 | 262 | 96 | ||
| GenBank mRNA | AF148447.1 | - | 2 - 3 | 205 | 96 | |
| AF175903.1 | - | 2 - 3 | 214 | 96 | ||
| AK007860.1 | - | 1 - 2 | 76 | 96 | ||
| AK011117.1 | - | 2 - 3 | 229 | 96 | ||
| AK017925.1 | - | 2 - 3 | 260 | 96 | ||
| BC006891.1 | - | 2 - 3 | 215 | 2650799 | 96 | |
| BY279731.1 | - | 2 - 3 | 262 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Uchl5
Genomic Map