Assay Details
Gene Symbol: Diap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019670.1 | NP_062644.1 | 9 - 10 | 952 | 62 | |
GenBank mRNA | AF094519.1 | - | 9 - 10 | 952 | 62 | |
AK137073.1 | - | 8 - 9 | 1041 | 62 | ||
AK138694.1 | - | 9 - 10 | 982 | 62 | ||
AK161737.1 | - | 8 - 9 | 970 | 62 | ||
AK166882.1 | - | 9 - 10 | 1008 | 62 | ||
AK220493.1 | - | 9 - 10 | 1016 | 62 | ||
BC003250.1 | - | 8 - 9 | 986 | 62 | ||
BC028920.1 | - | 2 - 3 | 253 | 4008236 | 62 | |
BC085191.1 | - | 9 - 10 | 1062 | 30693873 | 62 | |
BC086779.1 | - | 9 - 10 | 1027 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
