Assay Details
Gene Symbol: Diap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019670.1 | NP_062644.1 | 10 - 11 | 1060 | 106 | |
GenBank mRNA | AF094519.1 | - | 10 - 11 | 1060 | 106 | |
AK137073.1 | - | 9 - 10 | 1149 | 106 | ||
AK138694.1 | - | 10 - 11 | 1090 | 106 | ||
AK161737.1 | - | 9 - 10 | 1078 | 106 | ||
AK166882.1 | - | 10 - 11 | 1116 | 106 | ||
AK220493.1 | - | 10 - 11 | 1124 | 106 | ||
BC003250.1 | - | 9 - 10 | 1094 | 106 | ||
BC028920.1 | - | 3 - 4 | 361 | 4008236 | 106 | |
BC085191.1 | - | 10 - 11 | 1170 | 30693873 | 106 | |
BC086779.1 | - | 10 - 11 | 1135 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
