Assay Details
Gene Symbol: Diap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019670.1 | NP_062644.1 | 20 - 21 | 2301 | 57 | |
GenBank mRNA | AF094519.1 | - | 20 - 21 | 2301 | 57 | |
AK138694.1 | - | 20 - 21 | 2331 | 57 | ||
AK161737.1 | - | 19 - 20 | 2319 | 57 | ||
AK166882.1 | - | 20 - 21 | 2357 | 57 | ||
AK220493.1 | - | 20 - 21 | 2365 | 57 | ||
BC028920.1 | - | 13 - 14 | 1602 | 4008236 | 57 | |
BC085191.1 | - | 20 - 21 | 2411 | 30693873 | 57 | |
BC086779.1 | - | 20 - 21 | 2376 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
