Assay Details
Gene Symbol: Diap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019670.1 | NP_062644.1 | 22 - 23 | 2668 | 79 | |
GenBank mRNA | AF094519.1 | - | 22 - 23 | 2668 | 79 | |
AK138694.1 | - | 22 - 23 | 2698 | 79 | ||
AK161737.1 | - | 21 - 22 | 2686 | 79 | ||
AK166882.1 | - | 22 - 23 | 2724 | 79 | ||
AK220493.1 | - | 22 - 23 | 2732 | 79 | ||
BC028920.1 | - | 15 - 16 | 1969 | 4008236 | 79 | |
BC085191.1 | - | 22 - 23 | 2778 | 30693873 | 79 | |
BC086779.1 | - | 22 - 23 | 2743 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
