Assay Details
| Gene Symbol: Rbck1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083921.1 | NP_001077390.1 | 10 - 11 | 1677 | 78 | |
| NM_019705.3 | NP_062679.2 | 11 - 12 | 1518 | 78 | ||
| GenBank mRNA | AF124663.1 | - | 9 - 10 | 1287 | 78 | |
| AK085559.1 | - | 10 - 11 | 1666 | 79 | ||
| AK088591.1 | - | 10 - 11 | 1639 | 78 | ||
| AK146176.1 | - | 11 - 12 | 1692 | 78 | ||
| AK166075.1 | - | 10 - 11 | 1645 | 78 | ||
| AK199876.1 | - | 3 - 4 | 315 | 78 | ||
| BC032089.1 | - | 3 - 4 | 1122 | 78 | ||
| BC034555.1 | - | 11 - 12 | 1514 | 4192310 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rbck1
Genomic Map