Assay Details
| Gene Symbol: Serinc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019760.3 | NP_062734.1 | 6 - 7 | 990 | 114 | |
| GenBank mRNA | AB029500.1 | - | 6 - 7 | 934 | 114 | |
| AF181685.1 | - | 6 - 7 | 910 | 114 | ||
| AK002847.1 | - | 6 - 7 | 939 | 114 | ||
| AK005203.1 | - | 6 - 7 | 934 | 114 | ||
| AK088340.1 | - | 6 - 7 | 932 | 114 | ||
| AK133668.1 | - | 6 - 7 | 931 | 114 | ||
| AK133975.1 | - | 6 - 7 | 931 | 114 | ||
| AK135359.1 | - | 6 - 7 | 908 | 114 | ||
| AK141574.1 | - | 6 - 7 | 939 | 114 | ||
| AK148745.1 | - | 6 - 7 | 911 | 114 | ||
| AK159280.1 | - | 6 - 7 | 905 | 114 | ||
| AK159436.1 | - | 6 - 7 | 957 | 114 | ||
| AK167817.1 | - | 6 - 7 | 938 | 114 | ||
| BC006940.1 | - | 6 - 7 | 963 | 114 | ||
| BC017148.1 | - | 6 - 7 | 880 | 4506673 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Serinc1
Genomic Map