Assay Details
Gene Symbol: Abcb9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019875.2 | NP_063928.2 | 2 - 3 | 826 | 78 | |
GenBank mRNA | AB045382.2 | - | 1 - 2 | 646 | 78 | |
AB093298.1 | - | 2 - 3 | 808 | 78 | ||
AF216495.1 | - | 2 - 3 | 819 | 78 | ||
AK044140.1 | - | 2 - 3 | 804 | 78 | ||
BC053014.1 | - | 2 - 3 | 779 | 5686698 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
