Assay Details
Gene Symbol: Abcb9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019875.2 | NP_063928.2 | 3 - 4 | 945 | 69 | |
GenBank mRNA | AB045382.2 | - | 2 - 3 | 765 | 69 | |
AB093298.1 | - | 3 - 4 | 927 | 69 | ||
AF216495.1 | - | 3 - 4 | 938 | 69 | ||
AK044140.1 | - | 3 - 4 | 923 | 69 | ||
BC053014.1 | - | 3 - 4 | 898 | 5686698 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
