Assay Details
| Gene Symbol: Gfra4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136063.2 | NP_001129535.1 | 3 - 4 | 527 | 69 | |
| NM_001271001.1 | NP_001257930.1 | 3 - 4 | 527 | 69 | ||
| NM_001271002.1 | NP_001257931.1 | 3 - 4 | 554 | 69 | ||
| NM_020014.2 | NP_064398.1 | 3 - 4 | 554 | 69 | ||
| GenBank mRNA | AJ276514.1 | - | 3 - 4 | 554 | 69 | |
| AJ276516.1 | - | 3 - 4 | 554 | 69 | ||
| AJ276870.1 | - | 3 - 4 | 527 | 69 | ||
| AJ276871.1 | - | 3 - 4 | 527 | 69 | ||
| AK090192.1 | - | 1 - 2 | 191 | 69 | ||
| BC111808.1 | - | 3 - 4 | 553 | 69 | ||
| BC141873.1 | - | 3 - 4 | 549 | 69 | ||
| BC141874.1 | - | 3 - 4 | 554 | 40040202 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gfra4
Genomic Map