Assay Details
Gene Symbol: Med20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020048.3 | NP_064432.1 | 3 - 4 | 499 | 72 | |
GenBank mRNA | AJ245617.1 | - | 3 - 4 | 464 | 72 | |
AK043396.1 | - | 3 - 4 | 500 | 72 | ||
AK146345.1 | - | 3 - 4 | 500 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
protein ubiquitination
skeletal muscle cell differentiation
RNA polymerase II transcription cofactor activity
protein binding
ubiquitin protein ligase activity
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
protein ubiquitination
skeletal muscle cell differentiation
RNA polymerase II transcription cofactor activity
protein binding
ubiquitin protein ligase activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31213167 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P402319 | AK146345.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P402319 | BC060122.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P402319 | AK043396.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P402319 | NM_020048.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P402319 | AK003617.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P402319 | AK146345.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P402319 | BC060122.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P402319 | AK043396.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P402319 | NM_020048.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P402319 | AK003617.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 189874 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 154111 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 775448 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 57986 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 181099 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 828564 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 610128 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 33572 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 94800 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 57396 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P162648 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P107592 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P402319 | AK146345.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P402319 | BC060122.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P402319 | AK043396.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P402319 | NM_020048.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P402319 | AK003617.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P402319 | AK146345.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P402319 | BC060122.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P402319 | AK043396.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P402319 | NM_020048.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P402319 | AK003617.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P404067 | AJ245617.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31213181 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31213187 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31213172 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31213186 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31213185 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31213168 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31213189 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31213182 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31213170 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31213167 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31213176 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31213178 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31213174 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P162648 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P107592 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08792866 | - | - |
Whole Mouse Genome G4122A | A_52_P402319 | AK146345.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P402319 | BC060122.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P402319 | AK043396.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P402319 | NM_020048.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P402319 | AK003617.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P402319 | AK146345.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P402319 | BC060122.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P402319 | AK043396.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P402319 | NM_020048.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P402319 | AK003617.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P404067 | AJ245617.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101677 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132114 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P107592 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P162648 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13583790 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13583787 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13583801 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13262846 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31213167 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213292 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213237 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213176 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213265 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213235 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213231 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213304 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213247 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213167 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213228 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213267 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213190 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213170 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213202 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213258 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213262 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213168 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213233 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213174 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213209 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213242 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213195 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213259 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213307 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213249 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213255 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213179 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213287 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213271 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213283 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213207 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213196 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213229 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213206 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213273 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213281 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213254 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213285 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213201 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213276 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213290 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213193 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213246 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213239 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213269 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213298 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213248 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213172 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213175 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213189 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213278 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213251 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213244 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213305 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213181 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213275 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213210 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31213182 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13357984 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31213233 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31213234 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31213196 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13442398 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31213195 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13342754 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13583790 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31213189 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31213176 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31213179 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31213185 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31213190 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31213237 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31213199 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31213187 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31213170 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31213167 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13257657 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31213174 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31213172 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31213242 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31213239 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13183206 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31213178 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31213201 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31213181 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31213175 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31213235 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10445699 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10445696 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10445697 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10445700 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10445701 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10445698 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1416826_PM_a_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1448478_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10445695 | - | - |
Mouse Genome 430 2.0 Array | 1448478_at | - | - |
Mouse Genome 430 2.0 Array | 1439659_at | - | - |
Mouse Genome 430 2.0 Array | 1439659_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1416826_a_at | - | - |
Mouse Genome 430 2.0 Array | 1416826_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1448478_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56771_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56771_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1700008854.mm.1 | NM_020048.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700008854.mm.1 | NM_020048.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700008854.mm.1 | TC1700000921.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700008853.mm.1 | NM_020048.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700008853.mm.1 | NM_020048.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700008853.mm.1 | TC1700000921.mm.1 | MTA TC |
Mouse Expression Set 430 | 1448478_PM_at | - | - |
Mouse Expression Set 430 | 1416826_a_at | - | - |
Mouse Expression Set 430 | 1416826_PM_a_at | - | - |
Mouse Expression Set 430 | 1448478_at | - | - |
Mouse Expression Set 430 | 1439659_PM_at | - | - |
Mouse Expression Set 430 | 1439659_at | - | - |
Murine Genome U74v2 Set | 160724_at | - | - |
Murine Genome U74v2 Set | 160724_PM_at | - | - |
Murine Genome U74v2 Set | 162114_f_at | - | - |
Murine Genome U74v2 Set | 162114_PM_f_at | - | - |
Murine Genome U74v2 Set | 170302_r_at | - | - |
Murine Genome U74v2 Set | 167425_f_at | - | - |
Murine Genome U74v2 Set | 138571_at | - | - |
Murine Genome U74v2 Set | 170271_at | - | - |
Murine Genome U74v2 Set | 170302_PM_r_at | - | - |
Murine Genome U74v2 Set | 170271_PM_at | - | - |
Murine Genome U74v2 Set | 167425_PM_f_at | - | - |
Murine Genome U74v2 Set | 138571_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1448478_at | - | - |
Mouse Genome 430A 2.0 Array | 1416826_PM_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1416826_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1448478_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1448478_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1416826_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1416826_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1448478_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 102360722 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 102360722 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 104230609 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 70594 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000070594 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2467277 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1231001 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2467277 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1237521 | - | - |
mouse-6 v1.1 (Illumina) | scl056771.4_27-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 102360722 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 104230609 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 70594 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl056771.4_27-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 104230609 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 70594 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102360722 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2873729 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1237521 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1231001 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2873729 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl056771.4_27-S | - | - |
