Assay Details
| Gene Symbol: Mogs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020619.2 | NP_065644.2 | 3 - 4 | 846 | 58 | |
| GenBank mRNA | AK144946.1 | - | 3 - 4 | 847 | 58 | |
| AK157286.1 | - | 3 - 4 | 843 | 58 | ||
| BC051949.1 | - | 3 - 4 | 846 | 5716189 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591514 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591517 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591518 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P536481 | BC051949.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P536481 | AK152127.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P536481 | AK157286.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P536481 | AK219549.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P536481 | AK144946.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P536481 | BC051949.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P536481 | AK152127.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P536481 | AK157286.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P536481 | AK219549.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P536481 | AK144946.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 295407 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 503338 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 844894 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P536481 | BC051949.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P536481 | AK152127.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P536481 | AK157286.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P536481 | AK219549.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P536481 | AK144946.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P536481 | BC051949.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P536481 | AK152127.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P536481 | AK157286.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P536481 | AK219549.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P536481 | AK144946.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P188666 | - | - |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P536481 | BC051949.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P536481 | AK152127.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P536481 | AK157286.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P536481 | AK219549.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P536481 | AK144946.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P536481 | BC051949.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P536481 | AK152127.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P536481 | AK157286.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P536481 | AK219549.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P536481 | AK144946.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591515 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591530 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591512 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591529 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591506 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591499 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591503 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591514 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591528 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591509 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591498 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591533 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591524 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591510 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591496 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591494 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591501 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591535 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591520 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591518 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591523 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591505 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591493 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591522 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591526 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05960708 | - | - |
| Whole Mouse Genome G4122A | A_51_P188666 | - | - |
| Whole Mouse Genome G4122A | A_52_P536481 | BC051949.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P536481 | AK152127.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P536481 | AK157286.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P536481 | AK219549.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P536481 | AK144946.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P536481 | BC051949.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P536481 | AK152127.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P536481 | AK157286.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P536481 | AK219549.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P536481 | AK144946.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102511 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05759505 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05397485 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05612177 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591517 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591518 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591533 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591520 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591524 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591499 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05410551 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591542 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05303337 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05303306 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591512 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05397485 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591530 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05303318 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591493 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591508 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591511 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591507 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05303329 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05303319 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591535 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591523 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591518 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05303325 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591503 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05822286 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591496 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591498 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591522 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591533 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05600085 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10539338 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539340 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539341 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539339 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422489_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10539337 | - | - |
| Mouse Genome 430 2.0 Array | 1422489_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1422489_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 57377_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 57377_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600006954.mm.1 | TC0600000922.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600006954.mm.1 | NM_020619.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600006954.mm.1 | NM_020619.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600006955.mm.1 | TC0600000922.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600006955.mm.1 | NM_020619.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600006955.mm.1 | NM_020619.2 | Refseq |
| Mouse Expression Set 430 | 1422489_PM_at | - | - |
| Mouse Expression Set 430 | 1422489_at | - | - |
| Murine Genome U74v2 Set | 96139_PM_at | - | - |
| Murine Genome U74v2 Set | 96139_at | - | - |
| Murine Genome U74v2 Set | 168274_PM_f_at | - | - |
| Murine Genome U74v2 Set | 168274_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422489_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422489_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422489_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422489_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 393944 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1740603 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1740603 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001740603 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1255453 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1255453 | - | - |
| mouse-6 v1.1 (Illumina) | scl29846.4_459-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1740603 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl29846.4_459-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1740603 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1255453 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1255453 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl29846.4_459-S | - | - |

Mogs
Genomic Map