Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700000318.mm.1 |
TC0700000023.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700000318.mm.1 |
NM_001001454.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000318.mm.1 |
NM_001109765.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000318.mm.1 |
NM_021324.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000314.mm.1 |
TC0700000023.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700000314.mm.1 |
NM_001001454.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000314.mm.1 |
NM_001109765.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000314.mm.1 |
NM_021324.6 |
Refseq |