| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1500006993.mm.1 |
NM_021423.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500006993.mm.1 |
NM_021423.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500006993.mm.1 |
TC1500000878.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500007002.mm.1 |
NM_021423.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500007002.mm.1 |
NM_021423.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500007002.mm.1 |
TC1500000878.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500006997.mm.1 |
NM_021423.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500006997.mm.1 |
NM_021423.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500006997.mm.1 |
TC1500000878.mm.1 |
MTA TC |