Assay Details
Gene Symbol: Taf1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021441.2 | NP_067416.2 | 1 - 2 | 112 | 80 | |
GenBank mRNA | AK030522.1 | - | 1 - 2 | 79 | 80 | |
AK052250.1 | - | 1 - 2 | 79 | 80 | ||
AK087534.1 | - | 1 - 2 | 102 | 80 | ||
AK154846.1 | - | 1 - 2 | 96 | 80 | ||
AK156718.1 | - | 1 - 2 | 79 | 80 | ||
BC051944.1 | - | 1 - 2 | 91 | 80 | ||
BC058383.1 | - | 1 - 2 | 54 | 6811470 | 80 | |
Y09974.1 | - | 1 - 2 | 239 | 80 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK087534.1,AK154846.1,AK156718.1,BC058383.1,NM_021441.2,Y09974.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
