Assay Details
Gene Symbol: Taf1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021441.2 | NP_067416.2 | 10 - 11 | 1138 | 67 | |
GenBank mRNA | AK030522.1 | - | 10 - 11 | 1105 | 67 | |
AK052250.1 | - | 10 - 11 | 1178 | 67 | ||
AK087534.1 | - | 10 - 11 | 1128 | 67 | ||
AK154846.1 | - | 10 - 11 | 1122 | 67 | ||
BC051944.1 | - | 10 - 11 | 1190 | 67 | ||
BC058383.1 | - | 10 - 11 | 1080 | 6811470 | 67 | |
Y09974.1 | - | 10 - 11 | 1265 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
