Assay Details
Gene Symbol: Pmel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021882.4 | NP_068682.2 | 1 - 2 | 106 | 144 | |
GenBank mRNA | AK012808.1 | - | 1 - 2 | 107 | 144 | |
AK131956.1 | - | 1 - 2 | 108 | 144 | ||
BC075667.1 | - | 1 - 2 | 93 | 144 | ||
BC082555.1 | - | 1 - 2 | 92 | 30541702 | 144 | |
U14133.1 | - | 1 - 2 | 81 | 144 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
