Assay Details
Gene Symbol: Pmel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021882.4 | NP_068682.2 | 3 - 4 | 361 | 76 | |
GenBank mRNA | AK012808.1 | - | 3 - 4 | 362 | 76 | |
AK131956.1 | - | 3 - 4 | 363 | 76 | ||
AK204124.1 | - | 1 - 2 | 44 | 76 | ||
AK217723.1 | - | 1 - 2 | 36 | 76 | ||
BC075667.1 | - | 3 - 4 | 348 | 76 | ||
BC082555.1 | - | 3 - 4 | 347 | 30541702 | 76 | |
U14133.1 | - | 3 - 4 | 336 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P208145 | U14133.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P208145 | U14133.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 157964 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 475487 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 261769 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 694929 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 810376 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 170984 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 26071 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 276511 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 808636 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 212648 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 154027 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P135947 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P156226 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P208145 | U14133.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P208145 | U14133.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P208145 | U14133.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P208145 | U14133.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27542803 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27542796 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27542797 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27542800 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27542801 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27542805 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27542802 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27542810 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27542799 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27542809 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27542795 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P135947 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P156226 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07233902 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07233923 | - | - |
Whole Mouse Genome G4122A | A_51_P208145 | U14133.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P208145 | U14133.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101525 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136093 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P135947 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P156226 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08480050 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542765 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542797 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542821 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542839 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542755 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542788 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542746 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542830 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542740 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542775 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542759 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542749 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542859 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542916 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542826 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542914 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542828 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542773 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542744 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542748 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542781 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542796 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542774 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542860 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542809 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542793 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542761 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542911 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542732 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542856 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542833 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542787 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542837 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542851 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542777 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542753 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542913 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542784 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542905 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542863 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542799 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542854 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542769 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542918 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542861 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542846 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542794 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542823 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542848 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542843 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542907 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542801 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542733 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542819 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542795 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542779 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542742 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542751 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542785 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542813 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542758 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542831 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542776 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542862 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542864 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542804 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542803 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542817 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542799 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542791 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08480048 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542801 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542807 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542797 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542809 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542805 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08544185 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542793 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542815 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542814 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542816 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08208642 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542796 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10367384 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10367390 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10367389 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10367381 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10367391 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10367382 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10367388 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10367387 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10367385 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10367380 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10367392 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10367383 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10367386 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1422523_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10367379 | - | - |
Mouse Genome 430 2.0 Array | 1422523_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1422523_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20431_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20431_PM_at | - | - |
Affymetrix Murine Genome 6500A Array | Msa15640_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1000011892.mm.1 | NM_021882.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000011892.mm.1 | NM_021882.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000011892.mm.1 | TC1000001618.mm.1 | MTA TC |
Mouse Expression Set 430 | 1422523_at | - | - |
Mouse Expression Set 430 | 1422523_PM_at | - | - |
Murine Genome U74v2 Set | 102930_at | - | - |
Murine Genome U74v2 Set | 102930_PM_at | - | - |
Murine Genome U74v2 Set | 171583_PM_i_at | - | - |
Murine Genome U74v2 Set | 171583_i_at | - | - |
Mouse Genome 430A 2.0 Array | 1422523_at | - | - |
Mouse Genome 430A 2.0 Array | 1422523_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1422523_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1422523_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 839331 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 2260609 | - | - |
MouseWG-6 v2 Expression BeadChip | 730167 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2260609 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 730167 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000730167 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002260609 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2627503 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1230512 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1230512 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2627503 | - | - |
mouse-6 v1.1 (Illumina) | scl38274.14.1_69-S | - | - |
mouse-6 v1.1 (Illumina) | scl0003932.1_84-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2260609 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 730167 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl38274.14.1_69-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0003932.1_84-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2260609 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 730167 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1230512 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2627503 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1230512 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2627503 | - | - |
