Assay Details
Gene Symbol: Trpv6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022413.4 | NP_071858.3 | 3 - 4 | 645 | 82 | |
GenBank mRNA | AB037373.1 | - | 3 - 4 | 546 | 82 | |
AJ542487.1 | - | 3 - 4 | 350 | 82 | ||
AK077234.1 | - | 3 - 4 | 575 | 82 | ||
AK168760.1 | - | 3 - 4 | 645 | 82 | ||
BC016101.1 | - | 3 - 4 | 583 | 4911355 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
