Assay Details
Gene Symbol: Lat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020044.3 | NP_064428.1 | 8 - 9 | 500 | 73 | |
NM_022964.4 | NP_075253.2 | 7 - 8 | 464 | 73 | ||
GenBank mRNA | AF257136.1 | - | 7 - 8 | 444 | 73 | |
AK138953.1 | - | 8 - 9 | 459 | 73 | ||
AK143533.1 | - | 7 - 8 | 465 | 73 | ||
AK144261.1 | - | 8 - 9 | 439 | 73 | ||
AK162321.1 | - | 7 - 8 | 423 | 73 | ||
AY190024.1 | - | 8 - 9 | 465 | 73 | ||
BC005804.1 | - | 7 - 8 | 393 | 3154073 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
