Assay Details
| Gene Symbol: Lrp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022993.3 | NP_075369.2 | 3 - 4 | 641 | 57 | |
| GenBank mRNA | AB042200.1 | - | 3 - 4 | 543 | 57 | |
| AK132169.1 | - | 625 | 57 | |||
| AK144035.1 | - | 3 - 4 | 654 | 57 | ||
| AK170325.1 | - | 3 - 4 | 615 | 57 | ||
| AK185527.1 | - | 1 - 2 | 70 | 57 | ||
| BC052378.1 | - | 3 - 4 | 617 | 5696275 | 57 | |
| BC054447.1 | - | 3 - 4 | 609 | 6477698 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29612553 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29612546 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29612552 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29612551 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29612548 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29612545 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29612550 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29612549 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29612547 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P430357 | BC052378.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P430357 | BC054447.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P430357 | AK214260.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P430357 | AK214872.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P430357 | AK215152.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P430357 | BC052378.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P430357 | BC054447.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P430357 | AK214260.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P430357 | AK214872.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P430357 | AK215152.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 24826 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 851998 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 586141 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 909915 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 572957 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 63024 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P127435 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P430357 | BC052378.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P430357 | BC054447.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P430357 | AK214260.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P430357 | AK214872.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P430357 | AK215152.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P430357 | BC052378.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P430357 | BC054447.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P430357 | AK214260.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P430357 | AK214872.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P430357 | AK215152.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P430357 | BC052378.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P430357 | BC054447.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P430357 | AK214260.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P430357 | AK214872.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P430357 | AK215152.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P430357 | BC052378.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P430357 | BC054447.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P430357 | AK214260.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P430357 | AK214872.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P430357 | AK215152.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29612560 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29612561 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29612554 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29612537 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29612534 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29612551 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29612541 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29612540 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29612532 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29612549 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29612553 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29612545 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29612548 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29612563 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29612556 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29612539 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29612543 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02852026 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P127435 | - | - |
| Whole Mouse Genome G4122A | A_51_P430357 | BC052378.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P430357 | BC054447.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P430357 | AK214260.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P430357 | AK214872.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P430357 | AK215152.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P430357 | BC052378.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P430357 | BC054447.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P430357 | AK214260.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P430357 | AK214872.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P430357 | AK215152.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111029 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P127435 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11520930 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11431621 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11520928 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29612545 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29612552 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29612546 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29612549 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29612553 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29612550 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29612548 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29612551 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29612547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29612648 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11579114 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11520925 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29612585 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29612580 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29612579 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29612556 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29612532 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29612562 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29612590 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29612594 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29612560 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29612535 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11778736 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29612578 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11754409 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29612565 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29612568 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29612589 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29612540 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29612583 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29612551 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11778717 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29612587 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29612537 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29612538 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29612575 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29612591 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29612592 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29612534 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29612541 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29612554 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29612573 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29612564 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11754408 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29612533 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29612549 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29612586 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29612571 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29612569 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29612543 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11778751 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10415072 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10415070 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10415068 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10415067 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10415066 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10415069 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10415071 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10415073 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416836_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10415065 | - | - |
| Mouse Genome 430 2.0 Array | 1416836_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1416836_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 65107_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 65107_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1400005828.mm.1 | NM_022993.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400005828.mm.1 | NM_022993.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400005828.mm.1 | TC1400000747.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416836_at | - | - |
| Mouse Expression Set 430 | 1416836_PM_at | - | - |
| Murine Genome U74v2 Set | 161625_PM_r_at | - | - |
| Murine Genome U74v2 Set | 161625_r_at | - | - |
| Murine Genome U74v2 Set | 96186_PM_at | - | - |
| Murine Genome U74v2 Set | 96186_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416836_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416836_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416836_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416836_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 393745 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1580128 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001580128 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2760430 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2760430 | - | - |
| mouse-6 v1.1 (Illumina) | scl46303.7_279-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1580128 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl46303.7_279-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1580128 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2760430 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2760430 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl46303.7_279-S | - | - |

Lrp10
Genomic Map