Assay Details
| Gene Symbol: Cdc25b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111075.4 | NP_001104545.1 | 5 - 6 | 690 | 89 | |
| NM_023117.4 | NP_075606.1 | 5 - 6 | 690 | 89 | ||
| GenBank mRNA | AK004836.1 | - | 5 - 6 | 647 | 89 | |
| AK043730.1 | - | 5 - 6 | 681 | 89 | ||
| AK153903.1 | - | 5 - 6 | 691 | 89 | ||
| AK157397.1 | - | 5 - 6 | 648 | 89 | ||
| BC048164.1 | - | 5 - 6 | 687 | 89 | ||
| BC057568.1 | - | 5 - 6 | 561 | 6401489 | 89 | |
| BQ561430.1 | - | 5 - 6 | 346 | 89 | ||
| S93521.1 | - | 5 - 6 | 559 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdc25b
Genomic Map