Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00010791.mm.1 |
NM_001177955.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010791.mm.1 |
NM_001177955.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010791.mm.1 |
NM_001177956.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010791.mm.1 |
NM_001177956.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010791.mm.1 |
NM_001177957.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010791.mm.1 |
NM_001177957.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010791.mm.1 |
NM_001177958.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010791.mm.1 |
NM_001177958.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010791.mm.1 |
NM_001177959.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010791.mm.1 |
NM_001177959.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010791.mm.1 |
NM_001177960.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010791.mm.1 |
NM_001177960.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010791.mm.1 |
NM_001177961.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010791.mm.1 |
NM_001177961.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010791.mm.1 |
NM_001177962.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010791.mm.1 |
NM_001177962.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010791.mm.1 |
NM_023122.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010791.mm.1 |
NM_023122.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010791.mm.1 |
TC0X00001682.mm.1 |
MTA TC |