Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800017460.mm.1 |
NM_025436.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017460.mm.1 |
NM_025436.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017460.mm.1 |
TC0800002313.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800017466.mm.1 |
NM_025436.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017466.mm.1 |
NM_025436.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017466.mm.1 |
TC0800002313.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800017461.mm.1 |
NM_025436.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017461.mm.1 |
NM_025436.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017461.mm.1 |
TC0800002313.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800017468.mm.1 |
NM_025436.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017468.mm.1 |
NM_025436.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017468.mm.1 |
TC0800002313.mm.1 |
MTA TC |