Assay Details
| Gene Symbol: Dgat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026384.3 | NP_080660.1 | 2 - 3 | 451 | 84 | |
| GenBank mRNA | AF384160.1 | - | 2 - 3 | 309 | 84 | |
| AK002443.1 | - | 2 - 3 | 456 | 84 | ||
| AK016660.1 | - | 2 - 3 | 399 | 84 | ||
| BC043447.1 | - | 2 - 3 | 451 | 5125951 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dgat2
Genomic Map