Assay Details
| Gene Symbol: Cxxc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028868.3 | NP_083144.1 | 9 - 10 | 1485 | 76 | |
| GenBank mRNA | AK010337.1 | - | 9 - 10 | 1486 | 76 | |
| AK083655.1 | - | 9 - 10 | 1490 | 76 | ||
| AK146688.1 | - | 9 - 10 | 1569 | 76 | ||
| AK215977.1 | - | 1 - 2 | 62 | 76 | ||
| AY099096.1 | - | 9 - 10 | 1457 | 76 | ||
| BC012261.1 | - | 5 - 6 | 1028 | 76 | ||
| BC030938.1 | - | 9 - 10 | 1317 | 3980339 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cxxc1
Genomic Map