Assay Details
Gene Symbol: Kirrel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170985.1 | NP_001164456.1 | 4 - 5 | 813 | 108 | |
NM_130867.3 | NP_570937.2 | 4 - 5 | 810 | 108 | ||
GenBank mRNA | AF480411.1 | - | 3 - 4 | 448 | 108 | |
AK158381.1 | - | 4 - 5 | 767 | 108 | ||
AY017368.1 | - | 3 - 4 | 448 | 108 | ||
AY243095.1 | - | 3 - 4 | 448 | 108 | ||
BC023765.1 | - | 4 - 5 | 626 | 5343720 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
