Assay Details
Gene Symbol: Rogdi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133185.2 | NP_573448.2 | 5 - 6 | 380 | 62 | |
GenBank mRNA | AK002523.1 | - | 5 - 6 | 364 | 62 | |
AK046477.1 | - | 5 - 6 | 341 | 62 | ||
AK170144.1 | - | 5 - 6 | 345 | 62 | ||
AY036117.1 | - | 5 - 6 | 348 | 62 | ||
BC006914.1 | - | 5 - 6 | 423 | 3601096 | 62 | |
BU604779.1 | - | 5 - 6 | 397 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30513214 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30513219 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30513213 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30513217 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30513218 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30513215 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P303077 | BC006914.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P303077 | CD363552.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P303077 | NM_133185.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P303077 | AK002523.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P303077 | AK170144.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P303077 | BC006914.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P303077 | CD363552.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P303077 | NM_133185.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P303077 | AK002523.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P303077 | AK170144.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2111870 | BC006914.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2111870 | CD363552.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2111870 | NM_133185.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2111870 | AK046477.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2111870 | AY036117.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 97783 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 86094 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 1349 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2111870 | BC006914.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2111870 | CD363552.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2111870 | NM_133185.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2111870 | AK046477.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2111870 | AY036117.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P303077 | BC006914.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P303077 | CD363552.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P303077 | NM_133185.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P303077 | AK002523.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P303077 | AK170144.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P303077 | BC006914.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P303077 | CD363552.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P303077 | NM_133185.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P303077 | AK002523.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P303077 | AK170144.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P303077 | BC006914.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P303077 | CD363552.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P303077 | NM_133185.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P303077 | AK002523.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P303077 | AK170144.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P303077 | BC006914.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P303077 | CD363552.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P303077 | NM_133185.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P303077 | AK002523.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P303077 | AK170144.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P226788 | BC006914.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P226788 | CD363552.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P226788 | NM_133185.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P226788 | AK002523.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P226788 | AY036117.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30513202 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30513217 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30513198 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30513201 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30513200 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30513219 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30513205 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30513207 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30513209 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30513203 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30513215 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30513206 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30513208 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30513211 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30513210 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08493872 | - | - |
Whole Mouse Genome G4122A | A_52_P226788 | BC006914.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P226788 | CD363552.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P226788 | NM_133185.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P226788 | AK002523.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P226788 | AY036117.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P303077 | BC006914.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P303077 | CD363552.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P303077 | NM_133185.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P303077 | AK002523.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P303077 | AK170144.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P303077 | BC006914.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P303077 | CD363552.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P303077 | NM_133185.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P303077 | AK002523.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P303077 | AK170144.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102928 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12989636 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12732534 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30513215 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30513214 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30513219 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30513213 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30513217 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30513218 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30513207 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30513189 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30513195 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30513219 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30513185 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30513186 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30513190 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30513175 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30513206 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30513198 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30513191 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30513158 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30513179 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30513213 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30513210 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30513217 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30513155 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30513203 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30513200 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30513211 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30513176 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30513183 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30513194 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30513192 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30513215 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30513174 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30513208 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30513193 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30513205 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30513174 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30513201 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12563100 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30513207 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30513219 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30513189 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30513203 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30513183 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12989636 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30513190 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30513206 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30513198 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30513176 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30513191 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13021581 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30513179 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30513211 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12879150 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13021579 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13021583 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30513186 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12563085 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30513194 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30513208 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12670230 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30513213 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30513195 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10437488 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10437496 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10437486 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10437491 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10437485 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10437498 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10437487 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10437489 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10437492 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10437490 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10437497 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10437484 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10437493 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10437495 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1451421_PM_a_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10437483 | - | - |
Mouse Genome 430 2.0 Array | 1451421_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1451421_a_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66049_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66049_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1600009184.mm.1 | NM_133185.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600009184.mm.1 | NM_133185.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600009184.mm.1 | TC1600001150.mm.1 | MTA TC |
Mouse Expression Set 430 | 1451421_a_at | - | - |
Mouse Expression Set 430 | 1451421_PM_a_at | - | - |
Murine Genome U74v2 Set | 97228_at | - | - |
Murine Genome U74v2 Set | 97228_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1451421_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1451421_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1451421_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1451421_PM_a_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 536618 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 360020 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 360020 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000360020 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1222821 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1222821 | - | - |
mouse-6 v1.1 (Illumina) | scl066049.1_83-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 360020 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl066049.1_83-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 360020 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1222821 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1222821 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl066049.1_83-S | - | - |
