Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300012258.mm.1 |
NM_133222.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012258.mm.1 |
NM_133222.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012258.mm.1 |
TC0300001521.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300012260.mm.1 |
NM_133222.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012260.mm.1 |
NM_133222.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012260.mm.1 |
TC0300001521.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300012259.mm.1 |
NM_133222.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012259.mm.1 |
NM_133222.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012259.mm.1 |
TC0300001521.mm.1 |
MTA TC |